missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Beta-lactamase CTX-M-1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Beta-lactamase CTX-M-1, was expressed in the hosts and tags indicated.
2615.00 DKK - 23570.00 DKK
Specifications
Accession Number | WP_013188473.1 |
---|---|
Gene ID (Entrez) | 13909205 |
Name | Beta-lactamase CTX-M-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077510
|
enQuireBio™
QP7160-ye-10ug |
10 ÎĽg | N/A | ||||||
16017520
|
enQuireBio™
QP7160-ye-50ug |
50 ÎĽg | N/A | ||||||
16067510
|
enQuireBio™
QP7160-ye-100ug |
100 ÎĽg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16097510
|
enQuireBio™
QP7160-ye-200ug |
200 ÎĽg | N/A | ||||||
16007520
|
enQuireBio™
QP7160-ye-500ug |
500 ÎĽg |
19165.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16087510
|
enQuireBio™
QP7160-ye-1mg |
1 mg | N/A | ||||||
Specifications
WP_013188473.1 | |
Beta-lactamase CTX-M-1 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
13909205 | |
Research Use Only | |
bla | |
Recombinant Protein | |
Yeast | |
QTADVQQKLA ELERQSGGRL GVALINTADN SQILYRADER FAMCSTSKVM AVAAVLKKSE SEPNLLNQRV EIKKSDLVNY NPIAEKHVDG TMSLAELSAA ALQYSDNVAM NKLISHVGGP ASVTAFARQL GDETFRLDRT EPTLNTAIPG DPRDTTSPRA MAQTLRNLTL GKALGDSQRA QLVTWMKGNT TGAASIQAGL PASWVVGDKT GSGDYGTTND IAVIWPKDRA PLILVTYFTQ PQPKAESRRD VLASAAKIVT NGL | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Beta-lactamase CTX-M-1 Protein