missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecG Protein
A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase RecG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7112-200ug
This item is not returnable.
View return policy
Specifications
NP_418109.1 | |
ATP-dependent DNA helicase RecG Protein | |
Research Use Only | |
recG | |
Recombinant Protein | |
E. coli | |
MKGRLLDAVP LSSLTGVGAA LSNKLAKINL HTVQDLLLHL PLRYEDRTHL YPIGELLPGV YATVEGEVLN CNISFGGRRM MTCQISDGSG ILTMRFFNFS AAMKNSLAAG RRVLAYGEAK RGKYGAEMIH PEYRVQGDLS TPELQETLTP VYPTTEGVKQ ATLRKLTDQA LDLLDTCAIE ELLPPELSQG MMTLPEALRT LHRPPPTLQL SDLETGQHPA QRRLILEELL AHNLSMLALR AGAQRFHAQP LSANDTLKNK LLAALPFKPT GAQARVVAEI ERDMALDVPM MRLVQGDVGS GKTLVAALAA LRAIAHGKQV ALMAPTELLA EQHANNFRNW FAPLGIEVGW LAGKQKGKAR LAQQEAIASG QVQMIVGTHA IFQEQVQFNG LALVIIDEQH RFGVHQRLAL WEKGQQQGFH PHQLIMTATP IPRTLAMTAY ADLDTSVIDE LPPGRTPVTT VAIPDTRRTD IIDRVHHACI TEGRQAYWVC TLIEESELLE AQAAEATWEE LKLALPELNV GLVHGRMKPA EKQAVMASFK QGELHLLVAT TVIEVGVDVP NASLMIIENP ERLGLAQLHQ LRGRVGRGAV ASHCVLLYKT PLSKTAQIRL QVLRDSNDGF VIAQKDLEIR GPGELLGTRQ TGNAEFKVAD LLRDQAMIPE VQRLARHIHE RYPQQAKALI ERWMPETERY SNA | |
Greater than 90% as determined by SDS-PAGE. |
948162 | |
200 ÎĽg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecG Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction