missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L36 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L36, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8625-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005651

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417758.1
50S ribosomal protein L36 Protein
Research Use Only
rpmJ
Recombinant Protein
E. coli
MKVRASVKKL CRNCKIVKRD GVIRVICSAE PKHKQRQG
Greater than 90% as determined by SDS-PAGE.
32366016
50 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers