missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog PNLIPRP1 / PLRP1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) PNLIPRP1 / PLRP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6509-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003319.2 | |
PNLIPRP1 / PLRP1 Protein | |
Research Use Only | |
PNLIPRP1 | |
Recombinant Protein | |
E. coli | |
KEVCYEQIGC FSDAEPWAGT AIRPLKVLPW SPERIGTRFL LYTNKNPNNF QTLLPSDPST IEASNFQTDK KTRFIIHGFI DKGEENWLLD MCKNMFKVEE VNCICVDWKK GSQTSYTQAA NNVRVVGAQV AQMLSMLSAN YSYSPSQVQL IGHSLGAHVA GEAGSRTPGL GRITGLDPVE ASFQGTPEEV RLDPTDADFV DVIHTDAAPL IPFLGFGTSQ QMGHLDFFPN GGEEMPGCKK NALSQIVDLD GIWEGTRDFV ACNHLRSYKY YSESILNPDG FASYPCASYR AFESNKCFPC PDQGCPQMGH YADKFAVKTS DETQKYFLNT GDSSNFARWR YGVSITLSGK RATGQAKVAL FGSKGNTHQF NIFKGILKPG STHSNEFDAK LDVGTIEKVK FLWNNNVVNP TFPKVGAAKI TVQKGEEKTV HSFCSESTVR EDVLLTLTPC | |
Greater than 90% as determined by SDS-PAGE. |
404010 | |
200 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |