missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Viral interleukin-10 homolog Protein

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Viral interleukin-10 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7080-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087310

  • 2910.00 DKK / 10µg
Estimated Shipment: 31-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Viral interleukin-10 homolog Protein
10 μg
Not determined.
Full Length
Cytomegalovirus
His-SUMO
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UL111A
Recombinant Protein
E. coli
SEEAKPATTT IKNTKPQCRP EDYATRLQDL RVTFHRVKPT LQREDDYSVW LDGTVVKGCW GCSVMDWLLR RYLEIVFPAG DHVYPGLKTE LHSMRSTLES IYKDMRQCPL LGCGDKSVIS RLSQEAERKS DNGTRKGLSE LDTLFSRLEE YLHSRK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers