missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Envelope glycoprotein 1

A DNA sequence encoding the Human cytomegalovirus Envelope glycoprotein L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7456-ye-10ug

 View more versions of this product

Product Code. 16098900

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

YP_081555.1
Envelope glycoprotein 1
Research Use Only
gL
Recombinant Protein
Yeast
AAVSVAPTAA EKVPAECPEL TRRCLLGEVF EGDKYESWLR PLVNVTGRDG PLSQLIRYRP VTPEAANSVL LDEAFLDTLA LLYNNPDQLR ALLTLLSSDT APRWMTVMRG YSECGDGSPA VYTCVDDLCR GYDLTRLSYG RSIFTEHVLG FELVPPSLFN VVVAIRNEAT RTNRAVRLPV STAAAPEGIT LFYGLYNAVK EFCLRHQLDP PLLRHLDKYY AGLPPELKQT RVNLPAHSRY GPQAVDAR
Greater than 90% as determined by SDS-PAGE.
3077416
10 μg
Not determined.
Full Length
Cytomegalovirus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Cytomegalovirus Envelope glycoprotein 1 > 10μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.