missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cytomegalovirus Envelope glycoprotein 1
A DNA sequence encoding the Human cytomegalovirus Envelope glycoprotein L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7456-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
YP_081555.1 | |
Envelope glycoprotein 1 | |
Research Use Only | |
gL | |
Recombinant Protein | |
Yeast | |
AAVSVAPTAA EKVPAECPEL TRRCLLGEVF EGDKYESWLR PLVNVTGRDG PLSQLIRYRP VTPEAANSVL LDEAFLDTLA LLYNNPDQLR ALLTLLSSDT APRWMTVMRG YSECGDGSPA VYTCVDDLCR GYDLTRLSYG RSIFTEHVLG FELVPPSLFN VVVAIRNEAT RTNRAVRLPV STAAAPEGIT LFYGLYNAVK EFCLRHQLDP PLLRHLDKYY AGLPPELKQT RVNLPAHSRY GPQAVDAR | |
Greater than 90% as determined by SDS-PAGE. |
3077416 | |
50 μg | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |