Learn More
R&D Systems™ Recombinant Cynomolgus Monkey IgG1 Fc Protein
Product Code. 18795347
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity Unit Size
18795347 100 μg 100µg
1 options
Product Code. 18795347 Supplier R&D Systems™ Supplier No. 9315HG100
 Restricted Item
Forventet forsendelse: 27-11-2026
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Cynomolgus Monkey IgG1 Fc Protein is derived from HEK293. The Recombinant Cynomolgus Monkey IgG1 Fc Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Accession Number AAF14057
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 101867415
Molecular Weight (g/mol) 25 kDa
Name IgG1 Fc Chimera Recombinant Protein
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived cynomolgus monkey IgG1 protein Pro110-Lys333
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Common Name IgG1
Biological Activity 0.2 to 1.2ng/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
For Research Use Only.