missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Vitellogenin-2 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Vitellogenin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8859-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001026447.1 | |
Vitellogenin-2 Protein | |
Research Use Only | |
VTG2 | |
Recombinant Protein | |
E. coli | |
SRRSYLYNYE GSMLNGLQDR SLGKAGVRLS SKLEISGLPE NAYLLKVRSP QVEEYNGVWP RDPFTRSSKI TQVISSCFTR LFKFEYSSGR IGNIYAPEDC PDLCVNIVRG ILNMFQMTIK KSQNVYELQE AGIGGICHAR YVIQEDRKNS RIYVTRTVDL NNCQEKVQKS IGMAYIYPCP VDVMKERLTK GTTAFSYKLK QSDSGTLITD VSSRQVYQIS PFNEPTGVAV MEARQQLTLV EVRSERGSAP DVPMQNYGSL RYRFPAVLPQ MPLQLIKTKN PEQRIVETLQ HIVLNNQQDF HDDVSYRFLE VVQLCRIANA DNLESIWRQV SDKPRYRRWL LSAVSASGTT ETLKFLKNRI RNDDLNYIQT LLTVSLTLHL LQADEHTLPI AADLMTSSRI QKNPVLQQVA CLGYSSVVNR YCSQTSACPK EALQPIHDLA DEAISRGRED KMKLALKCIG NMGEPASLKR ILKFLPISSS SAADIPVHIQ IDAITALKKI AWKDPKTVQG YLIQILADQS LPPEVRMMAC AVIFETRPAL ALITTIANVA MKESNMQVAS FVYSHMKSLS KSRLPFMYNI SSACNIALKL LSPKLDSMSY RYSKVIRADT YFDNYRVGAT GEIFVVNS | |
Greater than 90% as determined by SDS-PAGE. |
424533 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Chicken Vitellogenin-2 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction