missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Focal adhesion kinase 1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Focal adhesion kinase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8800-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_990766.1 | |
Focal adhesion kinase 1 Protein | |
Research Use Only | |
PTK2 | |
Recombinant Protein | |
E. coli | |
LANNEKQGVR SHTVSVSETD DYAEIIDEED TYTMPSTRDY EIQRERIELG RCIGEGQFGD VHQGIYMSPE NPAMAVAIKT CKNCTSDSVR EKFLQEALTM RQFDHPHIVK LIGVITENPV WIIMELCTLG ELRSFLQVRK FSLDLASLIL YAYQLSTALA YLESKRFVHR DIAARNVLVS ATDCVKLGDF GLSRYMEDST YYKASKGKLP IKWMAPESIN FRRFTSASDV WMFGVCMWEI LMHGVKPFQG VKNNDVIGRI ENGERLPMPP NCPPTLYSLM TKCWAYDPSR RPRFTELKAQ LSTILEEE | |
Greater than 90% as determined by SDS-PAGE. |
396416 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
GST | |
Tris-based buffer, 50% glycerol |