missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Canine KGF/FGF-7 Protein
Klik for at se tilgængelige muligheder
Formulering:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
Beskrivelse
The Recombinant Canine KGF/FGF-7 Protein is derived from E. coli. The Recombinant Canine KGF/FGF-7 Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
| Gen-id (Entrez) | 403915 |
| Molekylvægt (g/mol) | 18.8 kDa |
| Navn | KGF/FGF-7 |
| Mængde | 25 μg |
| Kilde | E. coli-derived canine KGF/FGF-7 protein Cys32-Thr194 & Ser55-Thr194 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
R&D Systems™ Recombinant Canine KGF/FGF-7 Protein >
Ser du en mulighed for forbedring?Del en indholdskorrektion