missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Canine KGF/FGF-7 Protein
Klik for at se tilgængelige muligheder
Formulering:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
Beskrivelse
The Recombinant Canine KGF/FGF-7 Protein is derived from E. coli. The Recombinant Canine KGF/FGF-7 Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulering | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
| Gen-id (Entrez) | 403915 |
| Molekylvægt (g/mol) | 18.8 kDa |
| Navn | KGF/FGF-7 |
| Mængde | 25 μg |
| Kilde | E. coli-derived canine KGF/FGF-7 protein Cys32-Thr194 & Ser55-Thr194 |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FGF7, FGF-7, fibroblast growth factor 7HBGF-7, HBGF7, HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) |
| Konjugeret | Unconjugated |
| Rekombinant | Recombinant |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion