missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin, serovar A Protein
A DNA sequence encoding the Chlamydia trachomatis Major outer membrane porin, serovar A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7129-200ug
This item is not returnable.
View return policy
Specifications
Major outer membrane porin, serovar A Protein | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLKTDVNKEF QMGAAPTTRD VAGLEKDPVV NVARPNPAYG KHMQDAEMFT NAAYMALNIW DRFDVFCTLG ATTGYLKGNS ASFNLVGLFG TKTQSSGFDT ANIVPNTALN QAVVELYTDT TFAWSVGARA ALWECGCATL GASFQYAQSK PKVEELNVLC NASEFTINKP KGYVGAEFPL DITAGTEAAT GTKDASIDYH EWQASLALSY RLNMFTPYIG VKWSRVSFDA DTIRIAQPKL AKPVLDTTTL NPTIAGKGTV VSSAENELAD TMQIVSLQLN KMKSRKSCGI AVGTTIVDAD KYAVTVETRL IDERAAHVNA QFRF | |
Greater than 90% as determined by SDS-PAGE. |
200 ÎĽg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin, serovar A Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction