missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type A Protein

A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7051-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067200

  • 22245.00 DKK / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_003356619.1
1 mg
Not determined.
Partial (see sequence information for more details).
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
Botulinum neurotoxin type A Protein
Research Use Only
botA
Recombinant Protein
E. coli
MPFVNKQFNY KDPVNGVDIA YIKIPNVGQM QPVKAFKIHN KIWVIPERDT FTNPEEGDLN PPPEAKQVPV SYYDSTYLST DNEKDNYLKG VTKLFERIYS TDLGRMLLTS IVRGIPFWGG STIDTELKVI DTNCINVIQP DGSYRSEELN LVIIGPSADI IQFECKSFGH EVLNLTRNGY GSTQYIRFSP DFTFGFEESL EVDTNPLLGA GKFATDPAVT LAHELIHAGH RLYGIAINPN RVFKVNTNAY YEMSGLEVSF EELRTFGGHD AKFIDSLQEN EFRLYYYNKF KDIASTLNKA KSIVGTTASL QYMKNVFKEK YLLSEDTSGK FSVDKLKFDK LYKMLTEIYT EDNFVKFFKV LNRKTYLNFD KAVFKINIVP KVNYTIYDGF NLRNTNLAAN FNGQNTEINN MNFTKLKNFT GLFEFYKLLC VRGIIT
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers