missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Aspartate aminotransferase, mitochondrial Protein

A DNA sequence encoding the Bos taurus (Bovine) Aspartate aminotransferase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6108-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16080850

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_777231.1
Aspartate aminotransferase, mitochondrial Protein
Research Use Only
GOT2
Recombinant Protein
E. coli
GLAAAASARA SSWWAHVEMG PPDPILGVTE AFKRDTNSKK MNLGVGAYRD DNGKPYVLPS VRKAEAQIAA KNLDKEYLPI AGLAEFCKAS AELALGENNE VLKSGRYVTV QTISGTGALR IGASFLQRFF KFSRDVFLPK PTWGNHTPIF RDAGMQLQSY RYYDPKTCGF DFTGAIEDIS KIPAQSVILL HACAHNPTGV DPRPEQWKEM ATVVKKNNLF AFFDMAYQGF ASGDGNKDAW AVRHFIEQGI NVCLCQSYAK NMGLYGERVG AFTVVCKDAE EAKRVESQLK ILIRPMYSNP PINGARIAST ILTSPDLRKQ WLHEVKGMAD RIISMRTQLV SNLKKEGSSH NWQHIIDQIG MFCYTGLKPE QVERLTKEFS IYMTKDGRIS VAGVTSGNVA YLAHAIHQVT K
Greater than 90% as determined by SDS-PAGE.
286886
200 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers