missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Angiogenin-1 Protein
A DNA sequence encoding the Bos taurus (Bovine) Angiogenin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8810-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
Angiogenin-1 Protein | |
Research Use Only | |
ANG1 | |
Recombinant Protein | |
E. coli | |
AQDDYRYIHF LTQHYDAKPK GRNDEYCFNM MKNRRLTRPC KDRNTFIHGN KNDIKAICED RNGQPYRGDL RISKSEFQIT ICKHKGGSSR PPCRYGATED SRVIVVGCEN GLPVHFDESF ITPRH | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |