missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Single-stranded DNA-binding protein gp2.5 Protein

A DNA sequence encoding the Enterobacteria phage T7 (Bacteriophage T7) Single-stranded DNA-binding protein gp2.5, was expressed in the hosts and tags indicated.

3970.00 DKK - 24925.00 DKK

Specifications

Accession Number NP_041970.1
Gene ID (Entrez) 1261080
Name Single-stranded DNA-binding protein gp2.5 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16038591
View Documents
enQuireBio™
QP9695-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16078591
View Documents
enQuireBio™
QP9695-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028591
View Documents
enQuireBio™
QP9695-ye-100ug
100 ÎĽg
9855.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16058591
View Documents
enQuireBio™
QP9695-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068591
View Documents
enQuireBio™
QP9695-ye-500ug
500 ÎĽg
20720.00 DKK
500µg
Please to purchase this item. Need a web account? Register with us today!
16048591
View Documents
enQuireBio™
QP9695-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_041970.1
Single-stranded DNA-binding protein gp2.5 Protein
Not determined.
Full Length
Bacteriophage
His
Tris-based buffer, 50% glycerol
1261080
Research Use Only
2.5
Recombinant Protein
Yeast
MAKKIFTSAL GTAEPYAYIA KPDYGNEERG FGNPRGVYKV DLTIPNKDPR CQRMVDEIVK CHEEAYAAAV EEYEANPPAV ARGKKPLKPY EGDMPFFDNG DGTTTFKFKC YASFQDKKTK ETKHINLVVV DSKGKKMEDV PIIGGGSKLK VKYSLVPYKW NTAVGASVKL QLESVMLVEL ATFGGGEDDW ADEVEENGYV ASGSAKASKP RDEESWDEDD EESEEADEDG DF
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Bacteriophage Single-stranded DNA-binding protein gp2.5 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.