missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Bacteria Listeria monocytogenes p60 His Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP3-18146-10UG
Additional Details : Weight : 0.00970kg
Specifications
P21171 | |
SDS-PAGE | |
M.W. Theoretical: 50 kDa | |
Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. | |
IAP p60 from Listeria Monocytogenes, Invasion-associated Protein p60, Invasion-associated Protein p60 from Listeria Monocytogenes, invasion-associated-protein, p60 protein | |
>90% SDS-PAGE |
0.5 mg/ml | |
Liquid. 0.2um-filtered solution in PBS, pH 7.2 | |
10 μg | |
<1 EU/μg purified protein (LAL test; Lonza). | |
Immunology | |
Listeria monocytogenes p60 |