Learn More
enQuireBio™ Recombinant Bacillus Levanase Protein
A DNA sequence encoding the Bacillus sp. (strain L7) Levanase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7478-ec-50ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16098940
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Levanase Protein
Research Use Only
Partial (see sequence information for more details).
Bacillus
His-SUMO
Tris-based buffer, 50% glycerol
50 μg
Not determined.
Recombinant Protein
E. coli
LPWNDLGHVW SGSAVADTTN ASGLFGSSGG KGLIAYYTSY NPDRHNGNQK IGLAYSTDRG RTWKYSEEHP VVIENPGKTG EDPGGWDFRD PKVVRDEANN RWVMVVSGGD HIRLFTSTNL LNWTLTDQF
Greater than 90% as determined by SDS-PAGE.