missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. anthracis Single-stranded DNA-binding Protein
A DNA sequence encoding the Bacillus anthracis Single-stranded DNA-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7787-ec-100ug
This item is not returnable.
View return policy
Specifications
YP_031563.1 | |
Single-stranded DNA-binding Protein | |
Research Use Only | |
SSB | |
Recombinant Protein | |
E. coli | |
MNRVILVGRL TKDPDLRYTP NGVAVATFTL AVNRAFANQQ GEREADFINC VIWRKQAENV ANYLKKGSLA GVDGRLQTRN YEGQDGKRVY VTEVLAESVQ FLEPRNGGGE QRGSFNQQPS GAGFGNQSSN PFGQSSNSGN QGNQGNSGFT KNDDPFSNVG QPIDISDDDL PF | |
Greater than 95.4% as determined by SDS-PAGE. |
2848521 | |
100 ÎĽg | |
Not determined. | |
Full Length | |
B. anthracis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant B. anthracis Single-stranded DNA-binding Protein > 100μg
Spot an opportunity for improvement?Share a Content Correction