Learn More
enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein
A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7481-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
Modification methylase ApaLI Protein | |
100 μg | |
Not determined. | |
Full Length | |
A. pasturia | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
apaLIM | |
Recombinant Protein | |
E. coli | |
MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF |