missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein
A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7481-ye-50ug
This item is not returnable.
View return policy
Specifications
Modification methylase ApaLI Protein | |
Research Use Only | |
apaLIM | |
Recombinant Protein | |
Yeast | |
MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF | |
Greater than 90% as determined by SDS-PAGE. |
50 ÎĽg | |
Not determined. | |
Full Length | |
A. pasturia | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction