missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. pasturia Modification methylase ApaLI Protein

A DNA sequence encoding the Acetobacter pasteurianus (Acetobacter turbidans) Modification methylase ApaLI, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7481-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068960

  • 17300.00 DKK / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Modification methylase ApaLI Protein
Research Use Only
apaLIM
Recombinant Protein
Yeast
MNKDEVVVSL FAGAGGFSSG FSQAGLKPLF GAEINADACQ TYQENVGSPC HQLDLSTVDP SHIEMLTGGK RPFVVIGGPP CQGFSTAGPR NFADPRNLLI FNYLNIVERL SPRWLIFENV EGLLTSGGGR DLARLVREFV DMGYSVRLQK VNLAAYGVPQ TRKRVLIIGN RLGIDFQFPE ELYSFDSGKA KKASGKPLAP SLAEAVAGLG PAASDKDALV PYASSEPVNA FDARMRAGNR VEVVTHHVRV EAAERMQVEL LKPGQTMKDL PPELWHESYR RRANRRVSDG TPTEKRGGAP SGIKRLHGNL QSLTITGPAA REFIHPTEHR PLTIRECARI QTFPDKYRWV GNNASVIQQI GNAVPPLAAE RLAKHLRDID GSFGADTRPA GAMSAKLLGF VLTEALGMSP ALKSTEALLA EMHQGGFVF
Greater than 90% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length
A. pasturia
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers