Få mere at vide
Macherey-Nagel™ Receiver Plates
Artikelnummer. 16183547
Til brug med (applikation):
Centrifugation and Vacuum Use
Gravity Flow, Centrifugation, Vacuum Use
Inkluderer:
Filter Frits
Hydrophilised Filter Frits
Samlet volumen (metrisk) Well:
10 μL
35 μL
Pakningsstørrelse:
Pakke med 4
Artikelnummer. 16183547
Brand: Macherey-Nagel™ 740513.4
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Macherey-Nagel 96-well receiver plates with inserted filter frits of 35 μm pore size are suitable for centrifugation and vacuum applications for high throughput work.
  • 96-well plates
  • For centrifugation and vacuum
  • 35 μm filter frits
Tekniske data
Antal Wells 96
Samlet volumen (metrisk) Well 35 μL
Til brug med (applikation) Gravity Flow, Centrifugation, Vacuum Use
Inkluderer Hydrophilised Filter Frits