Få mere at vide
Reboxetine-d5 Mesylate, TRC
Artikelnummer. 30413238
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30413238 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413238 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1420022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Reboxetine mesylate-D5
CAS 1285918-53-7
Renhedsgrad noter HPLC
Molekylær formel C20H22D5NO6S
Mængde 2.5 mg
Synonym rel-(2R)-2-[(R)-[2-(Ethoxy-d5)phenoxy]phenylmethyl]morpholine methanesulfonate, Reboxetine-d5 mesylate, Reboxetine-D5 (O-ethyl-D5) mesilate
InChI formel InChI=1S/C19H23NO3.CH4O3S/c1-2-21-16-10-6-7-11-17(16)23-19(15-8-4-3-5-9-15)18-14-20-12-13-22-18;1-5(2,3)4/h3-11,18-20H,2,12-14H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1/i1D3,2D2;
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1ccccc1O[C@@H]([C@H]2CNCCO2)c3ccccc3.CS(=O)(=O)O
IUPAC navn methanesulfonic acid;(2R)-2-[(R)-[2-(1,1,2,2,2-pentadeuterioethoxy)phenoxy]-phenylmethyl]morpholine
Molekylvægt (g/mol) 414.53
Formel vægt 414.1873
Procent renhed >95