Få mere at vide
Rebaudioside C, TRC
Artikelnummer. 17987264
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17987264 5 mg
17929454 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17987264 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1395155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63550-99-2
Molekylær formel C44 H70 O22
Molekylvægt (g/mol) 951.01
Synonym Kaur-16-en-18-oic acid, 13-[(O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, β-D-glucopyranosyl ester, (4α)-, Dulcoside B, Rebaudioside C
SMIL C[C@@H]1O[C@@H](O[C@H]2[C@H](O[C@]34CC[C@H]5[C@]6(C)CCC[C@](C)([C@H]6CC[C@@]5(CC3=C)C4)C(=O)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)O[C@H](CO)[C@@H](O)[C@@H]2O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H](O)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Rebaudioside C
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C44H70O22/c1-17-12-43-10-6-22-41(3,8-5-9-42(22,4)40(58)65-38-33(57)30(54)26(50)20(14-46)61-38)23(43)7-11-44(17,16-43)66-39-35(64-36-31(55)28(52)24(48)18(2)59-36)34(27(51)21(15-47)62-39)63-37-32(56)29(53)25(49)19(13-45)60-37/h18-39,45-57H,1,5-16H2,2-4H3/t18-,19+,20+,21+,22-,23-,24-,25+,26+,27+,28+,29-,30-,31+,32+,33+,34-,35+,36-,37-,38-,39-,41+,42+,43+,44-/m0/s1
Formel vægt 950.4359
Procent renhed >95%