Få mere at vide
Rebamipide-d4, TRC
Artikelnummer. 30428278
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30428278 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428278 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1390021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Rebamipide-d4
CAS 1219409-06-9
Molekylær formel C19H11D4ClN2O4
Mængde 1 mg
Synonym α-[(4-Chlorobenzoyl-d4)-amino]-1,2-dihydro-2-oxo-4-quinolinepropanoic Acid, OPC-12759-d4, Mucosta-d4, Proamipide-d4
InChI formel InChI=1S/C19H15ClN2O4/c20-13-7-5-11(6-8-13)18(24)22-16(19(25)26)9-12-10-17(23)21-15-4-2-1-3-14(12)15/h1-8,10,16H,9H2,(H,21,23)(H,22,24)(H,25,26)/i5D,6D,7D,8D
SMIL O=C1NC2=CC=CC=C2C(CC(C(O)=O)NC(C3=C([2H])C([2H])=C(Cl)C([2H])=C3[2H])=O)=C1
Molekylvægt (g/mol) 374.81
Formel vægt 374.0971