missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ready-Made LB (Luria-Bertani) Broth Powder, Thermo Scientific Chemicals
For propagation of bacteria
367.82 DKK
Description
For the propagation of bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Off-white free flowing powder
- pH(2.0%, H2O): 7.0 ± 0.2
Contents
- Casein Peptone: 50%
- Yeast Extract: 25%
- Sodium Chloride: 25%
RUO – Research Use Only