missing translation for 'onlineSavingsMsg'
Få mere at vide

Reactive Yellow 86, Practical grade, MP Biomedicals™

Artikelnummer. 11455412 Shop alle MP Biomedicals produkter
Change view
Click to view available options
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
11455412 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11455412 Supplier MP Biomedicals Supplier No. 0219547925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale Reactive Yellow 86
CAS 61951-86-8
Molekylær formel C18H14Cl2N8Na2O9S2
InChI nøgle JUNRGNKPIKIXLL-UHFFFAOYSA-L
SMIL CCN1C(=O)C(=C(C(=NNC2=C(C=C(C(=C2)NC3=NC(=NC(=N3)Cl)Cl)S(=O)(=O)[O-])S(=O)(=O)[O-])C1=O)C)C(=O)N.[Na+].[Na+]
IUPAC navn disodium;4-[2-(5-carbamoyl-1-ethyl-4-methyl-2,6-dioxopyridin-3-ylidene)hydrazinyl]-6-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzene-1,3-disulfonate
Molekylvægt (g/mol) 667.357
PubChem CID 57369787
Grad Praktisk
Mængde 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.