missing translation for 'onlineSavingsMsg'
Få mere at vide

Reactive Violet 5 (~35%), TRC

Artikelnummer. 30416688
Change view
Click to view available options
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30416688 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416688 Supplier Toronto Research Chemicals Supplier No. TRCR12992525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Reactive Violet 5
CAS 12226-38-9
Molekylær formel C20 H16 N3 O15 S4 . 3 Na
Mængde 25 g
Synonym C.I. Reactive Violet 5 (8CI','9CI','ACI), 2,7-Naphthalenedisulfonic acid','5-(acetylamino)-4-hydroxy-3-[[2-hydroxy-4-[[2-(sulfooxy)ethyl]sulfonyl]phenyl]azo]-','copper complex (9CI), Copper','5-(acetylamino)-4-hydroxy-3-[[2-hydroxy-4-[[2-(sulfooxy)ethyl]sulfonyl]phenyl]azo]-2,7-naphthalenedisulfonic acid complex (9CI), Bezactiv Violet V 5R, Bezaktiv Violet V 5R, C.I. 18097, Cavalite Brilliant Violet 5R, Celmazol Brilliant Violet 5R, Diamira Brilliant Violet 5R, Intracron Brilliant Violet VS 5R, Primazin Brilliant Violet RL, Reactive Violet 5, Reactive Violet KN 4R, Reactofix Supra Violet H 5RL, Remazol Brilliant Violet 5R, Somazin Brilliant Violet 5R, Sunzol Brilliant Violet 5R, Sunzol Violet 5R
InChI formel InChI=1S/C20H19N3O15S4.3Na/c1-10(24)21-15-9-13(40(29,30)31)6-11-7-17(41(32,33)34)19(20(26)18(11)15)23-22-14-8-12(2-3-16(14)25)39(27,28)5-4-38-42(35,36)37;;;/h2-3,6-9,25-26H,4-5H2,1H3,(H,21,24)(H,29,30,31)(H,32,33,34)(H,35,36,37);;;/q;3*+1/p-3/b23-22+;;;
SMIL [Na+].[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(N=Nc3cc(ccc3O)S(=O)(=O)CCOS(=O)(=O)[O-])c(O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC navn trisodium;5-acetamido-4-hydroxy-3-[(E)-[2-hydroxy-5-(2-sulfonatooxyethylsulfonyl)phenyl]diazenyl]naphthalene-2,7-disulfonate
Molekylvægt (g/mol) 735.58
Formel vægt 734.9157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.