Learn More
Lenz Laborglasintrumente Reaction Vessels, Cylindrical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06007137
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11562204
  • 4815.00 DKK / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • With graduation
  • With laboratory flange LF
  • With groove
  • With round bottom
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
60 mm
0.1 L
185 mm