Learn More
Lenz Laborglasintrumente Reaction Vessels, Cylindrical
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06000149
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11502164
  • 1480.00 DKK / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • With graduation
  • With laboratory flange LF
  • With groove
  • With flat or round bottom
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
0.25 L
130 mm
Flat
Duran Glass
1 Ea.
65 mm