missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ RDBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577095
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
RDBP is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The protein encoded by this gene is part of a complex termed negative elongation factor (NELF) which represses RNA polymerase II transcript elongation. This protein bears similarity to nuclear RNA-binding proteins; however, it has not been demonstrated that this protein binds RNA. The protein contains a tract of alternating basic and acidic residues, largely arginine (R) and aspartic acid (D). The gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6.
Tekniske data
| RDBP | |
| Polyclonal | |
| Unconjugated | |
| NELFE | |
| D17H6S45; D6S45; major histocompatibility complex gene RD; negative elongation factor complex member E; negative elongation factor complex member E, Rdbp; negative elongation factor E; negative elongation factor polypeptide E; NELF E; Nelfe; NELF-E; nuclear protein; Rd; RD RNA binding protein; RD RNA-binding protein; RDBP; RDP; RNA-binding protein RD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27632, 294258, 7936 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18615, P19426 | |
| NELFE | |
| Recombinant full length Human RDBP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion