Få mere at vide
Tocris Bioscience™ RBC8
RalA and RalB GTPase inhibitor
Brand: Tocris Bioscience™ 5411/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
RBC8 is a RalA and RalB GTPase inhibitor (EC50 ∽3.5 μM). Inhibits the binding of Ral to RaLBP1. Exhibits no detectable inhibition of Ras or RhoA activity. Suppresses growth of xenograft tumors in mice. Cell permeable.Tekniske data
| Ras GTPase Inhibitors | |
| 6-Amino-4-(2,5-dimethoxyphenyl)-1,4-dihydro-3-(2-naphthalenyl)pyrano[2,3-c]pyrazole-5-carbonitrile | |
| Soluble to 100mM in DMSO | |
| Bioactive Small Molecule |
| 361185-42-4 | |
| 10 mg | |
| Solid |
Produkttitel
Ser du en mulighed for forbedring?
Research Use Only. Not For Human or Veterinary Use