missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Rat VEGF-165 Recombinant Protein, PeproTech®

Artikelnummer. 17804553
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17804553

Brand: Gibco™ 40031100UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Rat VEGF 165 is a 38.5 kDa, disulfide-linked homodimeric protein consisting of two 165 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P16612
Til brug med (applikation) ELISA, Functional Assay, Western Blot
Formulering protein with no preservative
Gen-id (Entrez) 83785
Molekylvægt (g/mol) 38.5 kDa
Navn Rat VEGF-165
Mængde 100 μg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <1 EU/ μg
Gene Alias H-VEGF-165; MVCD1; M-VEGF-165; vascular endothelial growth factor 165 b; vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; vascular permeability factor; VEGF; VEGF164; VEGF165; Vegfa; VEGF-A; VPF
Fælles navn VEGF-165
Gen symbol VEGFA
Biologisk aktivitet Determined by the dose-dependent stimulation of the proliferation of human umbilical vein endothelial cells (HUVEC). The expected ED50 is 4.0-8.0 ng/ml
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens MAPTTEGEQK AHEVVKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNVTMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.