missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Rat IL-13 (113 aa) Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Beskrivelse
Recombinant Rat IL-13 a 12.3 kDa protein consisting of 113 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
Tekniske data
Tekniske data
| Adgangsnummer | P42203 |
| Til brug med (applikation) | ELISA, Functional Assay, Western Blot |
| Formulering | protein with no preservative |
| Gen-id (Entrez) | 116553 |
| Molekylvægt (g/mol) | 12.3 kDa |
| Navn | Rat IL-13 (113 aa) |
| Mængde | 100 μg |
| Kilde | E. coli |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <1 EU/ μg |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion