Få mere at vide
R&D Systems™ Rat E-Selectin/CD62E (NP_620234.1) VersaClone cDNA
Artikelnummer. 18755638
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18755638 10 μg
1 options
Artikelnummer. 18755638 Leverandør R&D Systems™ Leverandørnr. RDC3001
  • ONLINE EXCLUSIVE PRICE
    2620.00 DKK
    2478.00 DKK / 10µg
    Spare 142.00 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 21-10-2026
Contains gene inserts that are fully sequenced and produced with high quality
TRUSTED_SUSTAINABILITY
Tekniske data
Gene E-Selectin/CD62E
Adgangsnummer NP_620234.1
Gen-id (Entrez) 6401
Gene Alias CD62 antigen-like family member E, CD62E, CD62E antigen, ELAM, ELAM1, ELAM-1, ELAM1E-selectin, endothelial adhesion molecule 1, Endothelial leukocyte adhesion molecule 1, ESEL, LECAM2, leukocyte endothelial cell adhesion molecule 2, Leukocyte-endothelial cell adhesion molecule 2, SELE, selectin E
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Rat
Produktlinje R&D Systems
Mængde 10 μg