Learn More
Novus Biologicals™ Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75860
This item is not returnable.
View return policy
Description
The Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric) measures rat Collagen III alpha 1/COL3A1 in serum, plasma and other biological fluids.
Specifications
0.63 - 40 ng/mL | |
This kit recognizes Rat COL3a1 in samples. No significant cross-reactivity or interference between Rat COL3a1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1281 | |
Rat | |
Cancer, Cell Biology, Extracellular Matrix | |
Collagen III alpha 1/COL3A1 |
0.38 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
COL3A1 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |