Learn More
Novus Biologicals™ Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75860
Product Code. 18310611
  • 6370.00 DKK / 1 set
Explore more special offers
Description
Description
The Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric) measures rat Collagen III alpha 1/COL3A1 in serum, plasma and other biological fluids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.63 - 40 ng/mL
This kit recognizes Rat COL3a1 in samples. No significant cross-reactivity or interference between Rat COL3a1 and analogues was observed.
Biological Fluids, Plasma, Serum
1281
Rat
Cancer, Cell Biology, Extracellular Matrix
Collagen III alpha 1/COL3A1
0.38 ng/mL
ELISA Kit (Colorimetric)
ELISA
COL3A1
1 Kit
Storage of components varies. See protocol for specific instructions.