Learn More
Novus Biologicals™ Rat Carboxypeptidase B2/CPB2 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand:  Novus Biologicals™ NBP3-06963
Product Code. 18310712
  • 6110.00 DKK / 1 set
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Specifications
1.56 to 100 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat Carboxypeptidase B2/CPB2 in samples. No significant cross-reactivity or interference between Rat Carboxypeptidase B2/CPB2 and analogues was observed.
1361
1 Kit
Storage of components varies. See protocol for specific instructions.
Rat
0.94 ng/mL
Cell Culture Supernatant, Plasma, Serum
ELISA, Sandwich ELISA
CPB2
Lipid and Metabolism
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor