Learn More
Novus Biologicals™ Rat Carboxypeptidase B2/CPB2 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Brand: Novus Biologicals™ NBP3-06963
This item is not returnable.
View return policy
Description
Specifications
1.56 to 100 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat Carboxypeptidase B2/CPB2 in samples. No significant cross-reactivity or interference between Rat Carboxypeptidase B2/CPB2 and analogues was observed. | |
1361 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Rat |
0.94 ng/mL | |
Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
CPB2 | |
Lipid and Metabolism | |
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor |