Få mere at vide
Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), DyLight 350, Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for ELISA
Tekniske data
Tekniske data
| Antigen | IgG2b Heavy Chain |
| Applikationer | ELISA |
| Klassifikation | Monoclonal |
| Klon | LO-MG2b-2 |
| Konjugeret | DyLight 350 |
| Formulering | 50mM Sodium Borate |
| Værtsarter | Rat |
| Immunogen | Purified mouse IgG2b |
| Oprensningsmetode | Protein A purified |
| Mængde | 0.25 mL |
| Vis mere |