Få mere at vide
Rapamycin Dialdehyde (Technical Grade), TRC
Artikelnummer. 30428525
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428525 10 mg
30415303 5 mg
30410328 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428525 Leverandør Toronto Research Chemicals Leverandørnr. TRCR12425010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500733-49-3
Molekylær formel C51H79NO15
Molekylvægt (g/mol) 946.17
Synonym (2S)-(2R,3S,6R,7E,9R,15E)-9-Hydroxy-1-((1S,4R)-4-hydroxy-3-methoxycyclohexyl)-10-methoxy-2,6,8,12,14-pentamethyl-5,11,17-trioxoheptadeca-7,15-dien-3-yl 1-(2-((2R,3R,6S)-2-hydroxy-6-((S,E)-2-methoxy-3-methyl-5-oxopent-3-en-1-yl)-3-methyltetrahydro-2H-pyran-2-yl)-2-oxoacetyl)piperidine-2-carboxylate
IUPAC navn [(2R,3S,6R,7E,9R)-9-hydroxy-1-[(1S,4R)-4-hydroxy-3-methoxycyclohexyl]-10-methoxy-2,6,8,12,14-pentamethyl-5,11,17-trioxoheptadeca-7,15-dien-3-yl] 1-[2-[(2R,3R,6S)-2-hydroxy-6-[(E,2S)-2-methoxy-3-methyl-5-oxopent-3-enyl]-3-methyloxan-2-yl]-2-oxoacetyl]piperidine-2-carboxylate
SMIL CO[C@@H](C[C@@H]1CC[C@@H](C)[C@@](O)(O1)C(=O)C(=O)N2CCCCC2C(=O)O[C@@H](CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)C(OC)C(=O)C(C)CC(C)C=CC=O)[C@H](C)C[C@@H]3CC[C@@H](O)C(C3)OC)\C(=C\C=O)\C
Tekniske data
Kemisk navn eller materiale Rapamycin Dialdehyde
Mængde 10 mg
InChI formel InChI=1S/C51H79NO15/c1-30(14-13-22-53)24-34(5)45(57)47(65-10)46(58)35(6)25-32(3)41(56)29-43(33(4)26-37-17-19-40(55)44(27-37)64-9)66-50(61)39-15-11-12-21-52(39)49(60)48(59)51(62)36(7)16-18-38(67-51)28-42(63-8)31(2)20-23-54/h13-14,20,22-23,25,30,32-34,36-40,42-44,46-47,55,58,62H,11-12,15-19,21,24,26-29H2,1-10H3/b14-13?,31-20+,35-25+/t30?,32-,33-,34?,36-,37+,38+,39?,40-,42+,43+,44?,46-,47?,51-/m1/s1
Formel vægt 945.545