Få mere at vide
Ranolazine USP Related Compound A, Mikromol™
Artikelnummer. 30051765
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051765 25 mg
30052539 100 mg
30459092 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051765 Leverandør Mikromol Leverandørnr. MM1320.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ranolazine USP Related Compound A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(2-Methoxyphenoxy)methyl]oxirane
CAS 2210-74-4
Analyt- eller komponentnavne 2-[(2-Methoxyphenoxy)methyl]oxirane
Forsendelsestilstand Room Temperature
Molekylær formel C10 H12 O3
Certificeringer/compliance ISO 17025
Synonym 2-[(2-Methoxyphenoxy)methyl]oxirane, Ranolazine Related Compound A
InChI formel InChI=1S/C10H12O3/c1-11-9-4-2-3-5-10(9)13-7-8-6-12-8/h2-5,8H,6-7H2,1H3
SMIL COc1ccccc1OCC2CO2
IUPAC navn 2-[(2-methoxyphenoxy)methyl]oxirane
Molekylvægt (g/mol) 180.2
Mængde 25 mg
Formel vægt 180.0786 g/mol