Få mere at vide
Tocris Bioscience™ Ranolazine dihydrochloride
Beskrivelse
Antianginal agent with antiarrhythmic properties that acts as a partial fatty acid oxidation inhibitor. Activates pyruvate dehydrogenase in ischemic myocytes to promote glucose oxidation, switching substrate utilization from fatty acids to glucose. Also shown to inhibit late INa and IKr currents.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ranolazine dihydrochloride |
| CAS | 95635-56-6 |
| Mængde | 50 mg |
| Molekylær formel | C24H35Cl2N3O4 |
| Formel vægt | 500.46 |
| Renhed | >99% |
| Molekylvægt (g/mol) | 500.46 |