Få mere at vide
Ranitidine N-Oxide, TRC
Artikelnummer. 30138698
Skift visning
Mængde:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138698 25 mg
17918414 10 mg
17977724 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138698 Leverandør Toronto Research Chemicals Leverandørnr. TRCR12001025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 73857-20-2
Molekylær formel C13 H22 N4 O4 S
Molekylvægt (g/mol) 330.4
Synonym 1,1-Ethenediamine, N-[2-[[[5-[(dimethyloxidoamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro- (9CI, ACI), 1,1-Ethenediamine, N-[2-[[[5-[(dimethylamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro-, N-oxide (ZCI), N-[2-[[[5-[(Dimethyloxidoamino)methyl]-2-furanyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine (ACI), N-[2-[[[5-[(Dimethyloxidoamino)-methyl]furan-2-yl]methyl]sulphanyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Ranitidine Hydrochloride Imp. E (EP), Ranitidine N-Oxide
IUPAC navn N,N-dimethyl-1-[5-[2-[[1-(methylamino)-2-nitroethenyl]amino]ethylsulfanylmethyl]furan-2-yl]methanamine oxide
SMIL CNC(=C[N+](=O)[O-])NCCSCc1oc(C[N+](C)(C)[O-])cc1
Tekniske data
Kemisk navn eller materiale Ranitidine N-Oxide
Mængde 25 mg
InChI formel InChI=1S/C13H22N4O4S/c1-14-13(8-16(18)19)15-6-7-22-10-12-5-4-11(21-12)9-17(2,3)20/h4-5,8,14-15H,6-7,9-10H2,1-3H3
Formel vægt 330.1362