Få mere at vide
Ranitidine hydrochloride, 99%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar B22260.14
| Mængde | 25g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAn H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.
Solubility
Soluble in water, 2-hydroxypropyl-beta-cyclodextrin, acetic acid, methanol, ethanol and dimethyl sulfoxide. Insoluble in chloroform.
Notes
Light sensitive. Incompatible with oxidizing agents.
Kemiske identifikatorer
| 66357-59-3 | |
| 350.862 | |
| GGWBHVILAJZWKJ-CHHCPSLASA-N | |
| 6603542 | |
| CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl |
| C13H23ClN4O3S | |
| MFCD00069339 | |
| ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
| (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride |
Tekniske data
| Ranitidine hydrochloride | |
| 99% | |
| MFCD00069339 | |
| 14,8110 | |
| Soluble in water,2-hydroxypropyl-beta-cyclodextrin,acetic acid,methanol,ethanol and dimethyl sulfoxide. Insoluble in chloroform. | |
| CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl | |
| 350.862 | |
| 350.86 |
| 66357-59-3 | |
| C13H23ClN4O3S | |
| 25g | |
| ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
| GGWBHVILAJZWKJ-CHHCPSLASA-N | |
| (Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride | |
| 6603542 | |
| 99% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.