Få mere at vide
Ranitidine EP Impurity H, Mikromol™
Artikelnummer. 30052268
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052268 100 mg
30051007 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052268 Leverandør Mikromol Leverandørnr. MM0086.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ranitidine EP Impurity H
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methyl-2-nitroacetamide
CAS 72078-82-1
Analyt- eller komponentnavne N-Methyl-2-nitroacetamide
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C3 H6 N2 O3
Certificeringer/compliance ISO 17025
Synonym Ranitidine Hydrochloride Imp. H (EP), Ranitidine Imp. H (EP), N-Methyl-2-nitroacetamide, Ranitidine Hydrochloride Impurity H, Ranitidine Impurity H
InChI formel InChI=1S/C3H6N2O3/c1-4-3(6)2-5(7)8/h2H2,1H3,(H,4,6)
SMIL CNC(=O)C[N+](=O)[O-]
IUPAC navn N-methyl-2-nitroacetamide
Molekylvægt (g/mol) 118.09
Mængde 100 mg
Formel vægt 118.0378 g/mol