Få mere at vide
Ranitidine EP Impurity G (Nizatidine EP Impurity K), Mikromol™
Artikelnummer. 30049555
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049555 100 mg
30051539 25 mg
30458407 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049555 Leverandør Mikromol Leverandørnr. MM0086.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ranitidine EP Impurity G (Nizatidine EP Impurity K)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(Methylamino)-5,6-dihydro-2H-1,4-thiazin-2-one Oxime
CAS 148639-72-9
Analyt- eller komponentnavne 3-(Methylamino)-5,6-dihydro-2H-1,4-thiazin-2-one Oxime
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C5 H9 N3 O S
Certificeringer/compliance ISO 17025
Synonym Nizatidine Imp. K (EP), Ranitidine Hydrochloride Imp. G (EP), Ranitidine Imp. G (EP), 3-(Methylamino)-5,6-dihydro-2H-1,4-thiazin-2-one Oxime, Nizatidine Impurity K, Ranitidine Hydrochloride Impurity G, Ranitidine Impurity G
InChI formel InChI=1S/C5H9N3OS/c1-6-4-5(8-9)10-3-2-7-4/h9H,2-3H2,1H3,(H,6,7)
SMIL CNC1=NCCSC1=NO
IUPAC navn N-[5-(methylamino)-2,3-dihydro-1,4-thiazin-6-ylidene]hydroxylamine
Molekylvægt (g/mol) 159.21
Mængde 100 mg
Formel vægt 159.0466 g/mol