missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ RANBP17 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15784101 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15784101 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15784101 Leverandør Invitrogen™ Leverandørnr. PA556193

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: MDGELSCRVF QLISLMDTGL PRCCNEKIEL AILWFLDQFR KTYVGDQLQR TSKVYARMSE VLGITDDNHV LETFMT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The transport of protein and large RNAs through the nuclear pore complexes (NPC) is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal (NLS) is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN (MIM 601179) plays a key role in NLS-dependent protein import. RAN-binding protein-17 is a member of the importin-beta superfamily of nuclear transport receptors.[supplied by OMIM, Jul 2002].
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen RANBP17
Applikationer Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RANBP17
Genadgangsnr. Q9H2T7
Gene Alias RAN binding protein 17; ran-binding protein 17; Ranbp17
Gensymboler RANBP17
Værtsarter Rabbit
Immunogen Recombinant protein corresponding to Human RANBP17. Recombinant protein control fragment (Product #RP-91863).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 64901
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.