missing translation for 'onlineSavingsMsg'
Få mere at vide

Ramosetron Hydrochloride, TRC

Artikelnummer. 30135657
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135657 100 mg
30136550 50 mg
30010679 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30135657 Leverandør Toronto Research Chemicals Leverandørnr. TRCR113050100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 132907-72-3
Molekylær formel C17 H17 N3 O . Cl H
Molekylvægt (g/mol) 315.8
Synonym (1-Methyl-1H-indol-3-yl)[(6R)-4,5,6,7-tetrahydro-1H-benzimidazol-6-yl]methanone Hydrochloride, (-)-(R)-5-[(1-Methyl-1H-indol-3-yl)carbonyl]-4,5,6,7-tetrahydro-1H- benzimidazole Monohydrochloride, (R)-(-)-Ramosetron Hydrochloride, Irribow, YM 060;
IUPAC navn (1-methylindol-3-yl)-[(5R)-4,5,6,7-tetrahydro-1H-benzimidazol-5-yl]methanone;hydrochloride
SMIL Cl.Cn1cc(C(=O)[C@@H]2CCc3[nH]cnc3C2)c4ccccc14

Tekniske data

Kemisk navn eller materiale Ramosetron Hydrochloride
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C17H17N3O.ClH/c1-20-9-13(12-4-2-3-5-16(12)20)17(21)11-6-7-14-15(8-11)19-10-18-14;/h2-5,9-11H,6-8H2,1H3,(H,18,19);1H/t11-;/m1./s1
Formel vægt 315.1138
Procent renhed >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.