Få mere at vide
Ramiprilat-d5 (Mixture of Diastereomers), TRC
Artikelnummer. 30414503
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30414503 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414503 Leverandør Toronto Research Chemicals Leverandørnr. TRCR1110121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ramiprilat phenyl-d5
CAS 1356837-92-7
Molekylær formel C21 2H5 H23 N2 O5
Mængde 1 mg
Synonym (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-Carboxy-3-(phenyl-d5)propyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic Acid, HOE 498 Diacid-d5, Ramipril Diacid-d5
InChI formel InChI=1S/C21H28N2O5/c1-13(22-16(20(25)26)11-10-14-6-3-2-4-7-14)19(24)23-17-9-5-8-15(17)12-18(23)21(27)28/h2-4,6-7,13,15-18,22H,5,8-12H2,1H3,(H,25,26)(H,27,28)/t13-,15+,16-,17+,18-/m0/s1/i2D,3D,4D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(CC[C@H](N[C@@H](C)C(=O)N2[C@@H]3CCC[C@@H]3C[C@H]2C(=O)O)C(=O)O)c([2H])c1[2H]
IUPAC navn (2S,3aR,6aR)-1-[(2S)-2-[[(1S)-1-carboxy-3-(2,3,4,5,6-pentadeuteriophenyl)propyl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
Molekylvægt (g/mol) 393.49
Formel vægt 393.2312